Anti-B4GALT1 Mouse Monoclonal Antibody [clone: 394706]

Supplier: R & D Systems
MAB3609
220020-540EA 594.37 USD
220020-540
Anti-B4GALT1 Mouse Monoclonal Antibody [clone: 394706]
Antibodies
Type: Primary
Antigen: B4GALT1
Clonality: Monoclonal
Clone: 394706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR