Anti-NPPB Rat Monoclonal Antibody [clone: 370124]

Supplier: R & D Systems
MAB36041
220020-539EA 594.37 USD
220020-539
Anti-NPPB Rat Monoclonal Antibody [clone: 370124]
Antibodies
Type: Primary
Antigen: NPPB
Clonality: Monoclonal
Clone: 370124
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR