Anti-HES4 Mouse Monoclonal Antibody [clone: 455237]

Supplier: R & D Systems
MAB3600
220020-537EA 577.02 USD
220020-537
Anti-HES4 Mouse Monoclonal Antibody [clone: 455237]
Antibodies
Type: Primary
Antigen: HES4
Clonality: Monoclonal
Clone: 455237
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR