Anti-ITGAL Mouse Monoclonal Antibody [clone: CR38]

Supplier: R & D Systems
MAB35951
102885-602EA 577.02 USD
102885-602
Anti-ITGAL Mouse Monoclonal Antibody [clone: CR38]
Antibodies
Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: CR38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR