Anti-SUMF2 Mouse Monoclonal Antibody [clone: 330312]

Supplier: R & D Systems
MAB3594
220020-533EA 561.11 USD
220020-533
Anti-SUMF2 Mouse Monoclonal Antibody [clone: 330312]
Antibodies
Type: Primary
Antigen: SUMF2
Clonality: Monoclonal
Clone: 330312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR