Anti-CD74 Mouse Monoclonal Antibody [clone: 332516]

Supplier: R & D Systems
MAB35901
102885-600EA 594.37 USD
102885-600
Anti-CD74 Mouse Monoclonal Antibody [clone: 332516]
Antibodies
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: 332516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR