Anti-EFNA3 Mouse Monoclonal Antibody [clone: 80814]

Supplier: R & D Systems
MAB359
220020-532EA 763.57 USD
220020-532
Anti-EFNA3 Mouse Monoclonal Antibody [clone: 80814]
Antibodies
Type: Primary
Antigen: EFNA3
Clonality: Monoclonal
Clone: 80814
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR