Anti-PPIA Rat Monoclonal Antibody [clone: 817815]

Supplier: R & D Systems
MAB3589
102885-598EA 594.37 USD
102885-598
Anti-PPIA Rat Monoclonal Antibody [clone: 817815]
Antibodies
Type: Primary
Antigen: PPIA
Clonality: Monoclonal
Clone: 817815
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR