Anti-CPE Mouse Monoclonal Antibody [clone: 420709]

Supplier: R & D Systems
MAB3587
220020-531EA 577.02 USD
220020-531
Anti-CPE Mouse Monoclonal Antibody [clone: 420709]
Antibodies
Type: Primary
Antigen: CPE
Clonality: Monoclonal
Clone: 420709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR