Anti-PRSS2 Mouse Monoclonal Antibody [clone: 410322]

Supplier: R & D Systems
MAB3586
220020-530EA 594.37 USD
220020-530
Anti-PRSS2 Mouse Monoclonal Antibody [clone: 410322]
Antibodies
Type: Primary
Antigen: PRSS2
Clonality: Monoclonal
Clone: 410322
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR