Anti-DSTYK Mouse Monoclonal Antibody [clone: 334640]

Supplier: R & D Systems
MAB3585
220020-529EA 647.88 USD
220020-529
Anti-DSTYK Mouse Monoclonal Antibody [clone: 334640]
Antibodies
Simple Western

Type: Primary
Antigen: DSTYK
Clonality: Monoclonal
Clone: 334640
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR