Anti-HSPA9 Mouse Monoclonal Antibody [clone: 419605]

Supplier: R & D Systems
MAB35841
220020-528EA 561.11 USD
220020-528
Anti-HSPA9 Mouse Monoclonal Antibody [clone: 419605]
Antibodies
Type: Primary
Antigen: HSPA9
Clonality: Monoclonal
Clone: 419605
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR