Anti-HSPA9 Mouse Monoclonal Antibody [clone: 419612]

Supplier: R & D Systems
MAB3584
220020-527EA 577.02 USD
220020-527
Anti-HSPA9 Mouse Monoclonal Antibody [clone: 419612]
Antibodies
Simple Western

Type: Primary
Antigen: HSPA9
Clonality: Monoclonal
Clone: 419612
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR