Anti-MET Mouse Monoclonal Antibody [clone: 95308]

Supplier: R & D Systems
MAB3583
220020-526EA 594.37 USD
220020-526
Anti-MET Mouse Monoclonal Antibody [clone: 95308]
Antibodies
Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: 95308
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR