Anti-FKBP8 Mouse Monoclonal Antibody [clone: 391505]

Supplier: R & D Systems
MAB3580
220020-523EA 594.37 USD
220020-523
Anti-FKBP8 Mouse Monoclonal Antibody [clone: 391505]
Antibodies
Type: Primary
Antigen Symbol: FKBP8
Antigen Name: FK506 binding protein 8, 38kDa
Antigen Synonyms: FKBP38, FKBPr38
Clonality: Monoclonal
Clone: 391505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR