Anti-MAP2K7 Mouse Monoclonal Antibody [clone: 324302]

Supplier: R & D Systems
MAB3579
220020-522EA 577.02 USD
220020-522
Anti-MAP2K7 Mouse Monoclonal Antibody [clone: 324302]
Antibodies
Type: Primary
Antigen: MAP2K7
Clonality: Monoclonal
Clone: 324302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR