Anti-DUSP6 Mouse Monoclonal Antibody [clone: 363246]

Supplier: R & D Systems
MAB3576
220020-520EA 577.02 USD
220020-520
Anti-DUSP6 Mouse Monoclonal Antibody [clone: 363246]
Antibodies
Type: Primary
Antigen: DUSP6
Clonality: Monoclonal
Clone: 363246
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR