Anti-FYN Mouse Monoclonal Antibody [clone: 361228]

Supplier: R & D Systems
MAB3574
220020-519EA 594.37 USD
220020-519
Anti-FYN Mouse Monoclonal Antibody [clone: 361228]
Antibodies
Type: Primary
Antigen Symbol: FYN
Antigen Name: FYN proto-oncogene, Src family tyrosine kinase
Antigen Synonyms: SLK, SYN, p59-FYN
Clonality: Monoclonal
Clone: 361228
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR