Anti-EGFR Mouse Monoclonal Antibody [clone: 338324]

Supplier: R & D Systems
MAB3570
220020-515EA 600.15 USD
220020-515
Anti-EGFR Mouse Monoclonal Antibody [clone: 338324]
Antibodies
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 338324
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR