Anti-KLK4 Mouse Monoclonal Antibody [clone: 325712]

Supplier: R & D Systems
MAB35661
220020-514EA 763.57 USD
220020-514
Anti-KLK4 Mouse Monoclonal Antibody [clone: 325712]
Antibodies
Type: Primary
Antigen: KLK4
Clonality: Monoclonal
Clone: 325712
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR