Anti-PRSS3 Rat Monoclonal Antibody [clone: 419910]

Supplier: R & D Systems
MAB35651
220020-513EA 561.11 USD
220020-513
Anti-PRSS3 Rat Monoclonal Antibody [clone: 419910]
Antibodies
Type: Primary
Antigen: PRSS3
Clonality: Monoclonal
Clone: 419910
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR