Anti-PRSS3 Rat Monoclonal Antibody [clone: 419911]

Supplier: R & D Systems
MAB3565
220020-512EA 577.02 USD
220020-512
Anti-PRSS3 Rat Monoclonal Antibody [clone: 419911]
Antibodies
Type: Primary
Antigen: PRSS3
Clonality: Monoclonal
Clone: 419911
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR