Anti-DDC Mouse Monoclonal Antibody [clone: 413911]

Supplier: R & D Systems
MAB3564
220020-511EA 594.37 USD
220020-511
Anti-DDC Mouse Monoclonal Antibody [clone: 413911]
Antibodies
Type: Primary
Antigen: DDC
Clonality: Monoclonal
Clone: 413911
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR