Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 94804]

Supplier: R & D Systems
MAB3561
220020-510EA 763.57 USD
220020-510
Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 94804]
Antibodies
Type: Primary
Antigen: TNFRSF14
Clonality: Monoclonal
Clone: 94804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR