Anti-CNDP2 Mouse Monoclonal Antibody [clone: 464702]

Supplier: R & D Systems
MAB3560
220020-509EA 577.02 USD
220020-509
Anti-CNDP2 Mouse Monoclonal Antibody [clone: 464702]
Antibodies
Type: Primary
Antigen: CNDP2
Clonality: Monoclonal
Clone: 464702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR