Anti-CHIT1 Mouse Monoclonal Antibody [clone: 397504]

Supplier: R & D Systems
MAB3559
220020-506EA 594.37 USD
220020-506
Anti-CHIT1 Mouse Monoclonal Antibody [clone: 397504]
Antibodies
Type: Primary
Antigen: CHIT1
Clonality: Monoclonal
Clone: 397504
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR