Anti-DISP1 Mouse Monoclonal Antibody [clone: 768213]

Supplier: R & D Systems
MAB3549
102885-596EA 561.11 USD
102885-596
Anti-DISP1 Mouse Monoclonal Antibody [clone: 768213]
Antibodies
Type: Primary
Antigen: DISP1
Clonality: Monoclonal
Clone: 768213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR