Anti-POSTN Rat Monoclonal Antibody [clone: 345613]

Supplier: R & D Systems
MAB3548
220020-502EA 600.15 USD
220020-502
Anti-POSTN Rat Monoclonal Antibody [clone: 345613]
Antibodies
Type: Primary
Antigen: POSTN
Clonality: Monoclonal
Clone: 345613
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR