Anti-SDC3 Rat Monoclonal Antibody [clone: 374412]

Supplier: R & D Systems
MAB35391
220020-499EA 577.02 USD
220020-499
Anti-SDC3 Rat Monoclonal Antibody [clone: 374412]
Antibodies
Type: Primary
Antigen: SDC3
Clonality: Monoclonal
Clone: 374412
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR