Anti-METAP1 Mouse Monoclonal Antibody [clone: 405419]

Supplier: R & D Systems
MAB3537
220020-497EA 577.02 USD
220020-497
Anti-METAP1 Mouse Monoclonal Antibody [clone: 405419]
Antibodies
Type: Primary
Antigen: METAP1
Clonality: Monoclonal
Clone: 405419
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR