Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 309707]

Supplier: R & D Systems
MAB3524
220020-496EA 561.11 USD
220020-496
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 309707]
Antibodies
Type: Primary
Antigen Symbol: NEUROG1
Antigen Name: neurogenin 1
Antigen Synonyms: AKA, Math4C, NEUROD3, bHLHa6, ngn1
Clonality: Monoclonal
Clone: 309707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR