Anti-MST1 Mouse Monoclonal Antibody [clone: 45904]

Supplier: R & D Systems
MAB352
220020-495EA 763.57 USD
220020-495
Anti-MST1 Mouse Monoclonal Antibody [clone: 45904]
Antibodies
Immunoaffinity Purification

Type: Primary
Antigen: MST1
Clonality: Monoclonal
Clone: 45904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR