Anti-IL12B Mouse Monoclonal Antibody [clone: 319429]

Supplier: R & D Systems
MAB3510
220020-493EA 763.57 USD
220020-493
Anti-IL12B Mouse Monoclonal Antibody [clone: 319429]
Antibodies
Type: Primary
Antigen: IL12B
Clonality: Monoclonal
Clone: 319429
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR