Anti-TM4SF18 Mouse Monoclonal Antibody [clone: 211116]

Supplier: R & D Systems
MAB3509
220020-492EA 577.02 USD
220020-492
Anti-TM4SF18 Mouse Monoclonal Antibody [clone: 211116]
Antibodies
Type: Primary
Antigen: TM4SF18
Clonality: Monoclonal
Clone: 211116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR