Anti-PTPRC Rat Monoclonal Antibody [clone: 319211]

Supplier: R & D Systems
MAB3507
220020-491EA 577.02 USD
220020-491
Anti-PTPRC Rat Monoclonal Antibody [clone: 319211]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 319211
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR