Anti-CK19 Mouse Monoclonal Antibody [clone: BA17]

Supplier: R & D Systems
MAB3506
220020-490EA 594.37 USD
220020-490
Anti-CK19 Mouse Monoclonal Antibody [clone: BA17]
Antibodies
Type: Primary
Antigen: CK19
Clonality: Monoclonal
Clone: BA17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR