Anti-TXNDC17 Mouse Monoclonal Antibody [clone: 544710]

Supplier: R & D Systems
MAB3504
220020-489EA 594.37 USD
220020-489
Anti-TXNDC17 Mouse Monoclonal Antibody [clone: 544710]
Antibodies
Type: Primary
Antigen: TXNDC17
Clonality: Monoclonal
Clone: 544710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR