Anti-ZBTB7A Mouse Monoclonal Antibody [clone: 466407]

Supplier: R & D Systems
MAB3496
220020-487EA 594.37 USD
220020-487
Anti-ZBTB7A Mouse Monoclonal Antibody [clone: 466407]
Antibodies
Type: Primary
Antigen: ZBTB7A
Clonality: Monoclonal
Clone: 466407
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR