Anti-LRIG3 Mouse Monoclonal Antibody [clone: 814834]

Supplier: R & D Systems
MAB3495
102885-592EA 594.37 USD
102885-592
Anti-LRIG3 Mouse Monoclonal Antibody [clone: 814834]
Antibodies
Type: Primary
Antigen: LRIG3
Clonality: Monoclonal
Clone: 814834
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR