Anti-MAPT Mouse Monoclonal Antibody [clone: 376720]

Supplier: R & D Systems
MAB3494
220020-486EA 594.37 USD
220020-486
Anti-MAPT Mouse Monoclonal Antibody [clone: 376720]
Antibodies
Type: Primary
Antigen: MAPT
Clonality: Monoclonal
Clone: 376720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR