Anti-PRDX2 Mouse Monoclonal Antibody [clone: 477719]

Supplier: R & D Systems
MAB3489
220020-482EA 594.37 USD
220020-482
Anti-PRDX2 Mouse Monoclonal Antibody [clone: 477719]
Antibodies
Type: Primary
Antigen: PRDX2
Clonality: Monoclonal
Clone: 477719
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR