Anti-PRDX1 Mouse Monoclonal Antibody [clone: 477609]

Supplier: R & D Systems
MAB3488
220020-481EA 633.42 USD
220020-481
Anti-PRDX1 Mouse Monoclonal Antibody [clone: 477609]
Antibodies
Type: Primary
Antigen: PRDX1
Clonality: Monoclonal
Clone: 477609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR