Anti-KLK9 Mouse Monoclonal Antibody [clone: 366721]

Supplier: R & D Systems
MAB3486
220020-480EA 577.02 USD
220020-480
Anti-KLK9 Mouse Monoclonal Antibody [clone: 366721]
Antibodies
Type: Primary
Antigen: KLK9
Clonality: Monoclonal
Clone: 366721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR