Anti-ERBB3 Mouse Monoclonal Antibody [clone: 526806]

Supplier: R & D Systems
MAB3483
220020-479EA 577.02 USD
220020-479
Anti-ERBB3 Mouse Monoclonal Antibody [clone: 526806]
Antibodies
Type: Primary
Antigen: ERBB3
Clonality: Monoclonal
Clone: 526806
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR