Anti-ERBB3 Mouse Monoclonal Antibody [clone: 526922]

Supplier: R & D Systems
MAB3482
220020-478EA 594.37 USD
220020-478
Anti-ERBB3 Mouse Monoclonal Antibody [clone: 526922]
Antibodies
Type: Primary
Antigen: ERBB3
Clonality: Monoclonal
Clone: 526922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR