Anti-CCR10 Rat Monoclonal Antibody [clone: 314305]

Supplier: R & D Systems
MAB3478
220020-474EA 623.29 USD
220020-474
Anti-CCR10 Rat Monoclonal Antibody [clone: 314305]
Antibodies
Type: Primary
Antigen: CCR10
Clonality: Monoclonal
Clone: 314305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR