Anti-CCR7 Rat Monoclonal Antibody [clone: 4B12]

Supplier: R & D Systems
MAB3477
220020-473EA 647.88 USD
220020-473
Anti-CCR7 Rat Monoclonal Antibody [clone: 4B12]
Antibodies
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR