Anti-RBP4 Rat Monoclonal Antibody [clone: 423614]

Supplier: R & D Systems
MAB34762
220020-472EA 577.02 USD
220020-472
Anti-RBP4 Rat Monoclonal Antibody [clone: 423614]
Antibodies
Type: Primary
Antigen: RBP4
Clonality: Monoclonal
Clone: 423614
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR