Anti-RBP4 Rat Monoclonal Antibody [clone: 423619]

Supplier: R & D Systems
MAB34761
102885-590EA 577.02 USD
102885-590
Anti-RBP4 Rat Monoclonal Antibody [clone: 423619]
Antibodies
Type: Primary
Antigen: RBP4
Clonality: Monoclonal
Clone: 423619
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR