Anti-METRN Rat Monoclonal Antibody [clone: 347511]

Supplier: R & D Systems
MAB34752
102885-588EA 763.57 USD
102885-588
Anti-METRN Rat Monoclonal Antibody [clone: 347511]
Antibodies
Type: Primary
Antigen: METRN
Clonality: Monoclonal
Clone: 347511
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR